3-Cyclopropyl-2-fluoropyridine 97%

Supplier: Ambeed
Warning

A109022-100MG A109022-250MG A109022-1G
76982-620EA 41 USD
76982-620 76982-622 77565-918
3-Cyclopropyl-2-fluoropyridine 97%
3-Cyclopropyl-2-fluoropyridine

Precaución: For research use only.

Formula: C₈H₈FN
MDL Number: MFCD20528324
CAS Number: 1227177-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR