3,5-Difluoro-2-nitroanisole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,5-Difluoro-3-methoxy-2-nitrobenzene, 3,5-Difluoro-2-nitrophenyl methyl ether
A108959-10G
A108959-1G
A108959-250MG
A108959-5G
A108959-100G
A108959-25G
76982-610EA
131.56
USD
76982-610
76982-612
76982-614
76982-616
77682-999
77683-000
3,5-Difluoro-2-nitroanisole 97%
3,5-Difluoro-2-nitroanisole
Precaución: For research use only.
Formula:
C₇H₅F₂NO₃ MW: 189.116 g/mol |
MDL Number:
MFCD11865203 CAS Number: 66684-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...