tert-Butyl-4-aminoazepane-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-aminoazepane-1-carboxylate

A108920-10G A108920-1G A108920-250MG A108920-25G A108920-5G
77008-816EA 234.46 USD
77008-816 77008-818 77008-820 77008-822 77008-824
tert-Butyl-4-aminoazepane-1-carboxylate 97%
tert-Butyl-4-aminoazepane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
MDL Number: MFCD07371372
CAS Number: 196613-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR