9-n-Octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 96%

Supplier: Ambeed
Warning

Synonyms: 9-Octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

A108833-1G A108833-250MG A108833-25G A108833-5G
76982-596EA 20.16 USD
76982-596 76982-598 76982-600 76982-602
9-n-Octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 96%
9-n-Octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole

Precaución: for Research Use Only

Formula: C₃₂H₄₇B₂NO₄
MW: 531.35 g/mol
MDL Number: MFCD23703114
CAS Number: 406726-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR