2-(4-Aminophenyl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Aminophenethyl alcohol

A108807-100G A108807-1G A108807-25G A108807-5G
77025-636EA 64.72 USD
77025-636 77025-638 77025-640 77025-642
2-(4-Aminophenyl)ethanol 98%
2-(4-Aminophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00007922
CAS Number: 104-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR