O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 97%

Supplier: Ambeed
Danger

A108798-10G A108798-25G
76995-090EA 25 USD
76995-090 76995-092
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 97%
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₂H₁₆BF₄N₅O₂
MDL Number: MFCD00077730
CAS Number: 125700-69-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR