3-Hydroxy-1-methacryloyloxyadamantane 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-1-adamantyl methacrylate, 3-Hydroxyadamantan-1-yl methacrylate, Methacrylic acid 3-Hydroxy-1-adamantyl ester

A108764-1G A108764-25G A108764-5G A108764-100G
77025-630EA 9 USD
77025-630 77025-632 77025-634 77570-504
3-Hydroxy-1-methacryloyloxyadamantane 97%
3-Hydroxy-1-methacryloyloxyadamantane

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
MW: 236.31 g/mol
MDL Number: MFCD13152044
CAS Number: 115372-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR