4,5-Dichloroquinoline 97%

Supplier: Ambeed
Warning

A108750-100MG A108750-1G A108750-250MG A108750-5G
77025-622EA 22 USD
77025-622 77025-624 77025-626 77025-628
4,5-Dichloroquinoline 97%
4,5-Dichloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
MDL Number: MFCD11858577
CAS Number: 21617-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR