2-Chloro-3,4-difluorobenzoic acid 95%

Supplier: Ambeed
Warning

A108677-1G A108677-250MG A108677-100MG
76982-592EA 49 USD
76982-592 76982-594 77889-757
2-Chloro-3,4-difluorobenzoic acid 95%
2-Chloro-3,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MDL Number: MFCD18415556
CAS Number: 150444-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR