2-Methoxy-6-nitrophenol 97%
Supplier: Ambeed
|
Warning
|
A108657-100MG
A108657-1G
A108657-250MG
A108657-5G
76982-584EA
12
USD
76982-584
76982-586
76982-588
76982-590
2-Methoxy-6-nitrophenol 97%
2-Methoxy-6-nitrophenol
Precaución: for Research Use Only
Formula:
C₇H₇NO₄ |
MDL Number:
MFCD08693430 CAS Number: 15969-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...