2-(9,9-Dioctyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A108647-1G A108647-250MG
77008-812EA 38 USD
77008-812 77008-814
2-(9,9-Dioctyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(9,9-Dioctyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₃₅H₅₃BO₂
MDL Number: MFCD20926226
CAS Number: 302554-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR