1,4-Bis(diphenylphosphino)butane 97%

Supplier: Ambeed
Warning

Synonyms: DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)

A108565-100G A108565-10G A108565-25G A108565-500G A108565-5G A108565-1000G
76995-052EA 46 USD
76995-052 76995-054 76995-056 76995-058 76995-060 77670-542
1,4-Bis(diphenylphosphino)butane 97%
1,4-Bis(diphenylphosphino)butane

Precaución: for Research Use Only

Formula: C₂₈H₂₈P₂
MDL Number: MFCD00003051
CAS Number: 7688-25-7
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR