2-(6-Bromopyridin-3-yl)pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A108509-100MG
A108509-1G
A108509-250MG
A108509-5G
77025-614EA
26.53
USD
77025-614
77025-616
77025-618
77025-620
2-(6-Bromopyridin-3-yl)pyrimidine 97%
2-(6-Bromopyridin-3-yl)pyrimidine
Precaución: for Research Use Only
Formula:
C₉H₆BrN₃ MW: 236.0680 g/mol |
MDL Number:
MFCD09999083 CAS Number: 942189-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...