(1S,4R)-Methyl 4-((tert-butoxycarbonyl)amino)cyclopent-2-enecarboxylate 97%
Supplier: Ambeed
|
Warning
|
A108492-1G
A108492-250MG
76982-580EA
6.37
USD
76982-580
76982-582
(1S,4R)-Methyl 4-((tert-butoxycarbonyl)amino)cyclopent-2-enecarboxylate 97%
(1S,4R)-Methyl 4-((tert-butoxycarbonyl)amino)cyclopent-2-enecarboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₉NO₄ MW: 241.28 g/mol |
MDL Number:
MFCD02259716 CAS Number: 168683-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...