S-Allyl-L-cysteine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-(Allylthio)-2-aminopropanoic acid, (R)-Allylthio-2-aminopropionic Acid

A108486-250MG A108486-1G
76995-040EA 8.49 USD
76995-040 77599-568
S-Allyl-L-cysteine 97%
S-Allyl-L-cysteine

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂S
MW: 161.22 g/mol
MDL Number: MFCD00151975
CAS Number: 21593-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR