N-Methyl-4-chlorobenzylamine Hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A108484-10G
A108484-1G
A108484-5G
76995-034EA
95
USD
76995-034
76995-036
76995-038
N-Methyl-4-chlorobenzylamine Hydrochloride 97%
N-Methyl-4-chlorobenzylamine Hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₁Cl₂N MW: 192.09 g/mol |
MDL Number:
MFCD07106759 CAS Number: 65542-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...