1,6-Dimethylphenanthro[1,2-b]furan-10,11-dione 97%

Supplier: Ambeed
Warning

A108483-100MG A108483-10MG A108483-1MG A108483-25MG A108483-50MG A108483-5MG A108483-1G
77025-602EA 53 USD
77025-602 77025-604 77025-606 77025-608 77025-610 77025-612 77871-427
1,6-Dimethylphenanthro[1,2-b]furan-10,11-dione 97%
1,6-Dimethylphenanthro[1,2-b]furan-10,11-dione

Precaución: for Research Use Only

Formula: C₁₈H₁₂O₃
MDL Number: MFCD00210563
CAS Number: 568-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR