Hexahydropyridazine dihydrochloride 97%

Supplier: Ambeed
Warning

A108468-1G A108468-250MG A108468-25G A108468-5G
77008-800EA 9 USD
77008-800 77008-802 77008-804 77570-492
Hexahydropyridazine dihydrochloride 97%
Hexahydropyridazine dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂Cl₂N₂
MW: 159.05 g/mol
MDL Number: MFCD09037903
CAS Number: 124072-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR