5-Bromo-2-methoxy-4-methylpyrimidine 98%

Supplier: Ambeed
Warning

A108464-100MG A108464-1G A108464-250MG A108464-5G
77008-792EA 28 USD
77008-792 77008-794 77008-796 77008-798
5-Bromo-2-methoxy-4-methylpyrimidine 98%
5-Bromo-2-methoxy-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MDL Number: MFCD18410559
CAS Number: 38696-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR