5-Cyclopropyl-2-fluoropyridine 97%

Supplier: Ambeed
Warning

A108426-250MG A108426-5G
76982-578EA 33 USD
76982-578 77889-756
5-Cyclopropyl-2-fluoropyridine 97%
5-Cyclopropyl-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₈H₈FN
MDL Number: MFCD18384289
CAS Number: 1034467-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR