Cyclohexyldiphenylphosphine oxide 98%

Supplier: Ambeed
Warning

A108388-100G A108388-10G A108388-1G A108388-25G A108388-5G
77025-592EA 352 USD
77025-592 77025-594 77025-596 77025-598 77025-600
Cyclohexyldiphenylphosphine oxide 98%
Cyclohexyldiphenylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₈H₂₁OP
MDL Number: MFCD00014301
CAS Number: 13689-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR