7-Chlorothieno[3,2-b]pyridine 98%

Supplier: Ambeed
Warning

A108387-100MG A108387-1G A108387-250MG A108387-25G A108387-5G
77008-776EA 5.31 USD
77008-776 77008-778 77008-780 77008-782 77008-784
7-Chlorothieno[3,2-b]pyridine 98%
7-Chlorothieno[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClNS
MDL Number: MFCD08448161
CAS Number: 69627-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR