Methyl 4,5-difluoro-2-nitro-benzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4,5-difluoro-2-nitrobenzoate

A108316-100G A108316-10G A108316-1G A108316-250MG A108316-5G A108316-25G
77025-576EA 384.05 USD
77025-576 77025-578 77025-580 77025-582 77025-584 77666-574
Methyl 4,5-difluoro-2-nitro-benzoate 97%
Methyl 4,5-difluoro-2-nitro-benzoate

Precaución: for Research Use Only

Formula: C₈H₅F₂NO₄
MW: 217.126 g/mol
MDL Number: MFCD11042825
CAS Number: 1015433-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR