3-Ethyl-4-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

A108311-10G A108311-1G A108311-250MG A108311-5G A108311-25G
76982-568EA 111.4 USD
76982-568 76982-570 76982-572 76982-574 77682-993
3-Ethyl-4-fluorobenzonitrile 97%
3-Ethyl-4-fluorobenzonitrile

Precaución: For research use only.

Formula: C₉H₈FN
MW: 149.165 g/mol
MDL Number: MFCD19441199
CAS Number: 869299-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR