1,3,5-Tris(4-carboxyphenyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 5'-(4-Carboxyphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid

A108224-1G A108224-250MG A108224-25G A108224-5G
77008-752EA 30.77 USD
77008-752 77008-754 77008-756 77008-758
1,3,5-Tris(4-carboxyphenyl)benzene 95%
1,3,5-Tris(4-carboxyphenyl)benzene

Precaución: for Research Use Only

Formula: C₂₇H₁₈O₆
MW: 438.44 g/mol
MDL Number: MFCD10000888
CAS Number: 50446-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR