N-Butanoyl-L-homoserine lactone 95%

Supplier: Ambeed
Warning

Synonyms: N-Butyryl-L-Homoserine Lactone, C4-HSL, (S)-N-(2-Oxotetrahydrofuran-3-yl)butyramide, N-(2-Oxotetrahydro-3-furanyl)butanamide, N-Butanoyl-HoSer·Lactone

A108181-100MG A108181-1G A108181-1MG A108181-250MG A108181-5G
76982-552EA 42.44 USD
76982-552 76982-554 76982-556 76982-558 77682-991
N-Butanoyl-L-homoserine lactone 95%
N-Butanoyl-L-homoserine lactone

Precaución: For research use only.

Formula: C₈H₁₃NO₃
MW: 171.2 g/mol
MDL Number: MFCD12912262
CAS Number: 67605-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR