L-Cysteine methyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Cys-OMe·HCl, (R)-Methyl 2-amino-3-mercaptopropanoate hydrochloride

A108035-10G A108035-1G A108035-5G A108035-100G A108035-25G A108035-1000G A108035-500G
76995-028EA 8 USD
76995-028 76995-030 76995-032 77565-896 77565-898 77599-562 77599-564
L-Cysteine methyl ester hydrochloride 98%
L-Cysteine methyl ester hydrochloride

Precaución: For research use only.

Formula: C₄H₉NO₂S·HCl
MW: 171.7 g/mol
MDL Number: MFCD00038998
CAS Number: 18598-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR