4-Chloro-5-iodo-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A108012-250MG A108012-1G A108012-5G A108012-10G
77025-564EA 35 USD
77025-564 77592-156 77592-158 77871-424
4-Chloro-5-iodo-1H-pyrrolo[2,3-b]pyridine 97%
4-Chloro-5-iodo-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClIN₂
MW: 278.48 g/mol
MDL Number: MFCD09965886
CAS Number: 1015610-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR