3-(Pyridin-4-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A107894-100MG A107894-1G A107894-250MG A107894-5G
77008-726EA 10 USD
77008-726 77008-728 77008-730 77008-732
3-(Pyridin-4-yl)benzonitrile 98%
3-(Pyridin-4-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₈N₂
MDL Number: MFCD06801941
CAS Number: 4350-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR