2-(Benzyloxy)-4-iodopyridine 98%

Supplier: Ambeed
Warning

A107877-100MG A107877-250MG
77008-722EA 63.66 USD
77008-722 77008-724
2-(Benzyloxy)-4-iodopyridine 98%
2-(Benzyloxy)-4-iodopyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀INO
MW: 311.12 g/mol
MDL Number: MFCD26401038
CAS Number: 896155-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR