Di-tert-butyl maleate 97%

Supplier: Ambeed
Warning

A107864-1G A107864-250MG
76982-520EA 93 USD
76982-520 76982-522
Di-tert-butyl maleate 97%
Di-tert-butyl maleate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
MDL Number: MFCD23381095
CAS Number: 18305-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR