Methyl 6-fluoro-1H-indole-4-carboxylate 97%

Supplier: Ambeed
Warning

A107863-100MG A107863-10G A107863-1G A107863-250MG A107863-5G
76982-510EA 25 USD
76982-510 76982-512 76982-514 76982-516 76982-518
Methyl 6-fluoro-1H-indole-4-carboxylate 97%
Methyl 6-fluoro-1H-indole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MDL Number: MFCD11845444
CAS Number: 1082040-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR