8-Bromo-7-methoxyquinoline 98%

Supplier: Ambeed
Warning

A107796-100MG A107796-1G A107796-250MG A107796-5G
77025-552EA 11 USD
77025-552 77025-554 77025-556 77025-558
8-Bromo-7-methoxyquinoline 98%
8-Bromo-7-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD16619426
CAS Number: 36023-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR