Ethyl 4-hydroxy-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carboxylate 95%

Supplier: Ambeed
Warning

A107770-100MG A107770-1G A107770-250MG
77008-712EA 82.76 USD
77008-712 77008-714 77008-716
Ethyl 4-hydroxy-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carboxylate 95%
Ethyl 4-hydroxy-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₄S
MW: 239.24 g/mol
MDL Number: MFCD22741552
CAS Number: 99429-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR