Ethyl-3-Amino-4-methylbenzoate 97%

Supplier: Ambeed
Warning

A107749-100G A107749-25G
77025-544EA 31 USD
77025-544 77025-546 77025-548
Ethyl-3-Amino-4-methylbenzoate 97%
Ethyl-3-Amino-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD01926538
CAS Number: 41191-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR