1-(4-Bromophenyl)pyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Bromophenyl)pyrrolidine

A107631-10G A107631-1G A107631-250MG A107631-25G A107631-5G
76995-012EA 123.07 USD
76995-012 76995-014 76995-016 76995-018 76995-020
1-(4-Bromophenyl)pyrrolidine 98%
1-(4-Bromophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN
MW: 226.1130 g/mol
MDL Number: MFCD04112480
CAS Number: 22090-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR