(4,6-Dimethylpyrimidin-2-yl)methanol 95%

Supplier: Ambeed
Warning

A107531-1G A107531-250MG
76982-496EA 259.93 USD
76982-496 76982-498
(4,6-Dimethylpyrimidin-2-yl)methanol 95%
(4,6-Dimethylpyrimidin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD16657415
CAS Number: 54198-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR