7-Bromo-4H-pyrido[1,2-a]pyrimidin-4-one 95%

Supplier: Ambeed
Warning

A107474-250MG A107474-50MG
76982-492EA 19 USD
76982-492 76982-494
7-Bromo-4H-pyrido[1,2-a]pyrimidin-4-one 95%
7-Bromo-4H-pyrido[1,2-a]pyrimidin-4-one

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MDL Number: MFCD07434965
CAS Number: 23536-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR