4,5,6,7-Tetrahydropyrazolo[1,5-a]pyrimidine 95%

Supplier: Ambeed
Warning

A107431-100MG A107431-250MG A107431-1G A107431-5G
77008-708EA 29 USD
77008-708 77008-710 77592-152 77889-744
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyrimidine 95%
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MDL Number: MFCD08273904
CAS Number: 126352-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR