7,8-Difluorochroman-4-one 97%

Supplier: Ambeed
Warning

A107407-1G A107407-250MG
77025-524EA 193.09 USD
77025-524 77025-526
7,8-Difluorochroman-4-one 97%
7,8-Difluorochroman-4-one

Precaución: for Research Use Only

Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
MDL Number: MFCD11518476
CAS Number: 890840-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR