(3-Bromo-2-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

A107233-10G A107233-1G A107233-250MG A107233-25G A107233-5G
77025-508EA 42 USD
77025-508 77025-510 77025-512 77025-514 77025-516
(3-Bromo-2-pyridinyl)methanol 97%
(3-Bromo-2-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MDL Number: MFCD03095348
CAS Number: 52378-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR