4-Bromo-3-methoxybenzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

A107203-250MG A107203-5G A107203-1G
77008-688EA 24 USD
77008-688 77008-690 77592-150
4-Bromo-3-methoxybenzenesulfonyl chloride 95%
4-Bromo-3-methoxybenzenesulfonyl chloride
Formula: C₇H₆BrClO₃S
MDL Number: MFCD14702923
CAS Number: 1215295-91-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR