4,4'-(Hexafluoroisopropylidene)diphenol 98%
Supplier: Ambeed
|
Warning
|
A107192-100G
A107192-10G
A107192-25G
A107192-500G
A107192-5G
76994-988EA
35.01
USD
76994-988
76994-990
76994-992
76994-994
76994-996
4,4'-(Hexafluoroisopropylidene)diphenol 98%
4,4'-(Hexafluoroisopropylidene)diphenol
Precaución: for Research Use Only
Formula:
C₁₅H₁₀F₆O₂ |
MDL Number:
MFCD00000439 CAS Number: 1478-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...