Ethyl 4-cyclopropyl-2,4-dioxobutanoate 97%

Supplier: Ambeed
Warning

A107168-1G A107168-250MG A107168-25G A107168-5G
76982-480EA 46.68 USD
76982-480 76982-482 76982-484 76982-486
Ethyl 4-cyclopropyl-2,4-dioxobutanoate 97%
Ethyl 4-cyclopropyl-2,4-dioxobutanoate

Precaución: for Research Use Only

Formula: C₉H₁₂O₄
MW: 184.18 g/mol
MDL Number: MFCD09052462
CAS Number: 21080-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR