5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Formyl-2,4-dimethyl-3-pyrrolecarboxylic acid, 5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid
A107049-100G
A107049-10G
A107049-1G
A107049-25G
A107049-500G
A107049-5G
76994-976EA
176.11
USD
76994-976
76994-978
76994-980
76994-982
76994-984
76994-986
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 98%
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C8H9NO3 MW: 167.16 g/mol |
MDL Number:
MFCD06202342 CAS Number: 253870-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...