(1,2-Dimethyl-1H-benzo[d]imidazol-5-yl)methanol 98%

Supplier: Ambeed
Warning

A106986-100MG A106986-1G A106986-5G
76982-468EA 47.75 USD
76982-468 76982-470 76982-472
(1,2-Dimethyl-1H-benzo[d]imidazol-5-yl)methanol 98%
(1,2-Dimethyl-1H-benzo[d]imidazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.21 g/mol
MDL Number: MFCD02690737
CAS Number: 4589-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR