4-Bromo-5-methylisoxazole 97%

Supplier: Ambeed
Warning

A106951-100MG A106951-1G A106951-250MG
77025-498EA 67.9 USD
77025-498 77025-500 77025-502
4-Bromo-5-methylisoxazole 97%
4-Bromo-5-methylisoxazole

Precaución: for Research Use Only

Formula: C₄H₄BrNO
MW: 161.985 g/mol
MDL Number: MFCD13689016
CAS Number: 7064-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR