1-Cyclopentyl-4-methylbenzene 97%

Supplier: Ambeed
Warning

A106816-10G A106816-250MG
77008-660EA 442.4 USD
77008-660 77008-662
1-Cyclopentyl-4-methylbenzene 97%
1-Cyclopentyl-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆
MW: 160.26 g/mol
MDL Number: MFCD11111744
CAS Number: 827-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR