5-Butyl-1H-benzotriazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Butyl-1H-benzo[d][1,2,3]triazole

A106814-10G A106814-1G A106814-25G A106814-5G
77008-652EA 367.08 USD
77008-652 77008-654 77008-656 77008-658
5-Butyl-1H-benzotriazole 98%
5-Butyl-1H-benzotriazole

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃
MW: 175.23 g/mol
MDL Number: MFCD22581692
CAS Number: 3663-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR