(3-Bromo-5-fluorophenyl)methanamine 95%

Supplier: Ambeed
Danger

A106759-100MG A106759-250MG
77025-490EA 47.75 USD
77025-490 77025-492
(3-Bromo-5-fluorophenyl)methanamine 95%
(3-Bromo-5-fluorophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.03 g/mol
MDL Number: MFCD11619398
CAS Number: 1094555-68-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR